Tisk…

Azetidine hydrochloride 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: Azetidine HCl

OR59858-1G OR59858-25G OR59858-5G
APOSOR59858-1GEA 1410 CZK
APOSOR59858-1G APOSOR59858-25G APOSOR59858-5G
Azetidine hydrochloride 95%
Azetidine hydrochloride
Vzorec: C₃H₈ClN
MW: 93,56 g/mol
Bod tání: 300 °C
Bod vzplanutí: 152 °C
Storage Temperature: Ambient
MDL Number: MFCD00191762
CAS číslo: 36520-39-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR