2,5-Dichlorobenzoyl chloride
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR59857-1G
OR59857-250MG
APOSOR59857-1GEA
705
CZK
APOSOR59857-1G
APOSOR59857-250MG
2,5-Dichlorobenzoyl chloride
2,5-Dichlorobenzoyl chloride
Vzorec:
C₇H₃Cl₃O MW: 209,46 g/mol Bod varu: 95 °C/11,2mmHg Bod tání: 23 °C Storage Temperature: Ambient |
MDL Number:
MFCD00035821 CAS číslo: 2905-61-5 EINECS: 220-812-0 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...