4-Hydrazinoquinoline hydrochloride
Dodavatel: Apollo Scientific
|
Varování
|
OR59842-1G
OR59842-250MG
OR59842-5G
APOSOR59842-1GEA
10350
CZK
APOSOR59842-1G
APOSOR59842-250MG
APOSOR59842-5G
4-Hydrazinoquinoline hydrochloride
4-Hydrazinoquinoline hydrochloride
Vzorec:
C₉H₁₀ClN₃ Storage Temperature: Ambient |
MDL Number:
MFCD01861876 CAS číslo: 68500-41-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...