Tisk…

2-Amino-6-methoxypyridine

Dodavatel: Apollo Scientific
Varování

OR5984-1G OR5984-25G OR5984-5G
APOSOR5984-1GEA 622 CZK
APOSOR5984-1G APOSOR5984-25G APOSOR5984-5G
2-Amino-6-methoxypyridine
2-Amino-6-methoxypyridine
Vzorec: C₆H₈N₂O
MW: 124,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04972542
CAS číslo: 17920-35-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR