Tisk…

2-Amino-5-bromo-3-iodopyridine 96%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 5-Bromo-3-iodo-2-pyridylamine, 5-Bromo-3-iodopyridin-2-amine

OR59815-25G
APOSOR59815-25GEA 1740 CZK
APOSOR59815-25G
2-Amino-5-bromo-3-iodopyridine 96%
2-Amino-5-bromo-3-iodopyridine
Vzorec: C₅H₄BrIN₂
MW: 298,91 g/mol
Bod tání: 112…115 °C
Storage Temperature: Ambient
MDL Number: MFCD06659000
CAS číslo: 381233-96-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR