Tisk…

4-Bromo-2-methoxybenzoic acid

Dodavatel: Apollo Scientific
Nebezpečí

OR59811-1G OR59811-5G OR59811-100G
APOSOR59811-1GEA 622 CZK
APOSOR59811-1G APOSOR59811-5G APOSOR59811-100G
4-Bromo-2-methoxybenzoic acid
4-Bromo-2-methoxybenzoic acid
Vzorec: C₈H₇BrO₃
Bod tání: 155…159 °C
Storage Temperature: Ambient
MDL Number: MFCD07780665
CAS číslo: 72135-36-5
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR