Tisk…

Methyl-3-{[(2-methoxy-2-oxoethyl)amino]sulphonyl}thiophene-2-carboxylate

Dodavatel: Apollo Scientific

Synonyms: Methyl 3-(N-(2-methoxy-2-oxoethyl)sulphamoyl)thiophene-2-carboxylate

OR5979-1G OR5979-5G
APOSOR5979-1GEA 417 CZK
APOSOR5979-1G APOSOR5979-5G
Methyl-3-{[(2-methoxy-2-oxoethyl)amino]sulphonyl}thiophene-2-carboxylate
Methyl-3-{[(2-methoxy-2-oxoethyl)amino]sulphonyl}thiophene-2-carboxylate
Vzorec: C₉H₁₁NO₆S₂
MW: 293,32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01570165
CAS číslo: 106820-63-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR