Tisk…

7-Indoleboronic acid pinacol ester 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 7-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole

OR59787-1G OR59787-5G
APOSOR59787-1GEA 662 CZK
APOSOR59787-1G APOSOR59787-5G
7-Indoleboronic acid pinacol ester 97%
7-Indoleboronic acid pinacol ester
Vzorec: C₁₄H₁₈BNO₂
MW: 243,1092 g/mol
Bod tání: 83…85 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08669591
CAS číslo: 642494-37-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR