Chloromethanesulphonyl chloride 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR5975-5G
OR5975-1G
APOSOR5975-5GEA
2860
CZK
APOSOR5975-5G
APOSOR5975-1G
Chloromethanesulphonyl chloride 95%
Chloromethanesulphonyl chloride
Vzorec:
CH₂Cl₂O₂S Bod varu: 175 °C Hustota: 1,67 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00014727 CAS číslo: 3518-65-8 EINECS: 222-526-1 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...