Tisk…

5-Nitro-8-quinolinol 98%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 5-Nitroquinolin-8-ol, Nitroxoline, 8-Hydroxy-5-nitroquinoline

OR5948-100G OR5948-25G
APOSOR5948-100GEA 3310 CZK
APOSOR5948-100G APOSOR5948-25G
5-Nitro-8-quinolinol 98%
5-Nitro-8-quinolinol
Vzorec: C₉H₆N₂O₃
MW: 190,16 g/mol
Bod tání: 181…183 °C
Storage Temperature: Ambient
MDL Number: MFCD00006791
CAS číslo: 4008-48-4
EINECS: 223-662-4
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR