Methyl-2-(3-aminophenyl)acetate hydrochloride
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Methyl 2-(3-aminophenyl)acetate hydrochloride
OR59469-1G
OR59469-5G
OR59469-25G
APOSOR59469-1GEA
827
CZK
APOSOR59469-1G
APOSOR59469-5G
APOSOR59469-25G
Methyl-2-(3-aminophenyl)acetate hydrochloride
Methyl-2-(3-aminophenyl)acetate hydrochloride
Vzorec:
C₉H₁₂ClNO₂ MW: 201.6501 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11501071 CAS číslo: 150319-83-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...