Methyl-4-(chloromethyl)benzoate
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
Methyl 4-(chloromethyl)benzoate
OR59468-100G
OR59468-10G
OR59468-25G
APOSOR59468-100GEA
10780
CZK
APOSOR59468-100G
APOSOR59468-10G
APOSOR59468-25G
Methyl-4-(chloromethyl)benzoate
Methyl-4-(chloromethyl)benzoate
Vzorec:
C₉H₉ClO₂ MW: 184,62 g/mol Bod varu: 160…165 °C/30mmHg Bod tání: 37…40 °C Storage Temperature: Ambient |
MDL Number:
MFCD00511224 CAS číslo: 34040-64-7 EINECS: 251-806-6 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...