Methyl (diethylphosphono)acetate
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Diethyl methoxycarbonylmethanephosphonate, Methyl 2-(diethoxyphosphoryl)acetate, Methyl diethylphosphonoacetate
OR59465-100G
OR59465-1G
OR59465-10G
OR59465-50G
APOSOR59465-100GEA
1870
CZK
APOSOR59465-100G
APOSOR59465-1G
APOSOR59465-10G
APOSOR59465-50G
Methyl (diethylphosphono)acetate
Methyl (diethylphosphono)acetate
Vzorec:
C₇H₁₅O₅P MW: 210.17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00009081 CAS číslo: 1067-74-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...