Tisk…

4-Chloro-3-methylphenyl isocyanate 97%

Dodavatel: Apollo Scientific
Nebezpečí

OR59456-1G OR59456-5G
APOSOR59456-1GEA 3410 CZK
APOSOR59456-1G APOSOR59456-5G
4-Chloro-3-methylphenyl isocyanate 97%
4-Chloro-3-methylphenyl isocyanate

  • May form small amount of sediment.

Vzorec: C₈H₆ClNO
MW: 167,60 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11101408
CAS číslo: 51488-20-1
UN: 2206
ADR: 6.1,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR