4-Chloro-2-isocyanato-1-methylbenzene 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
5-Chloro-2-methylphenyl isocyanate, 5-Chloro-2-methylphenylisocyanate
OR59455-100G
OR59455-25G
OR59455-5G
OR59455-1G
APOSOR59455-100GEA
0
CZK
APOSOR59455-100G
APOSOR59455-25G
APOSOR59455-5G
APOSOR59455-1G
4-Chloro-2-isocyanato-1-methylbenzene 98%
4-Chloro-2-isocyanato-1-methylbenzene
- May form small amount of sediment.
Vzorec:
C₈H₆ClNO Bod varu: 115 °C/5mmHg Hustota: 1,226 g/cm³ (25 °C) Bod vzplanutí: 108 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00019913 CAS číslo: 40411-27-6 UN: 2206 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...