Tisk…

3-Methoxyanthranilic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Amino-m-anisic acid, 3-Methoxyanthranilic acid, o-Anisidine-6-carboxylic acid, 2-Amino-3-methoxybenzoic acid

OR59444-10G OR59444-1G OR59444-5G
APOSOR59444-10GEA 1410 CZK
APOSOR59444-10G APOSOR59444-1G APOSOR59444-5G
3-Methoxyanthranilic acid
3-Methoxyanthranilic acid
Vzorec: C₈H₉NO₃
MW: 167,16 g/mol
Bod tání: 169…172 °C
Storage Temperature: Ambient
MDL Number: MFCD00075178
CAS číslo: 3177-80-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR