Diethyl acetylenedicarboxylate 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR59420-100G
OR59420-25G
APOSOR59420-100GEA
2240
CZK
APOSOR59420-100G
APOSOR59420-25G
Diethyl acetylenedicarboxylate 95%
Diethyl acetylenedicarboxylate
Vzorec:
C₈H₁₀O₄ Bod varu: 107…110 °C/11mmHg Bod tání: 44228 °C Hustota: 1,063 g/cm³ (25 °C) Bod vzplanutí: 94 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00009186 CAS číslo: 762-21-0 EINECS: 212-095-8 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...