3-Bromo-N-cyclohexylbenzamide
Dodavatel: Apollo Scientific
OR59407-1G
OR59407-5G
APOSOR59407-1GEA
3740
CZK
APOSOR59407-1G
APOSOR59407-5G
3-Bromo-N-cyclohexylbenzamide
3-Bromo-N-cyclohexylbenzamide
Vzorec:
C₁₃H₁₆BrNO Bod tání: 166…168 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00554081 CAS číslo: 59507-55-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...