6-Chloro-1-methyl-2-indoleboronic acid 96%
Dodavatel: Apollo Scientific
|
Varování
|
OR59404-1G
OR59404-250MG
OR59404-100MG
APOSOR59404-1GEA
17000
CZK
APOSOR59404-1G
APOSOR59404-250MG
APOSOR59404-100MG
6-Chloro-1-methyl-2-indoleboronic acid 96%
6-Chloro-1-methyl-2-indoleboronic acid
Vzorec:
C₉H₉BClNO₂ Bod tání: 118…120 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09878352 CAS číslo: 957066-11-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...