Tisk…

N-Ethyl-N-(4-methoxybenzyl)benzenesulphonamide

Dodavatel: Apollo Scientific

OR59397-1G OR59397-5G
APOSOR59397-1GEA 2070 CZK
APOSOR59397-1G APOSOR59397-5G
N-Ethyl-N-(4-methoxybenzyl)benzenesulphonamide
N-Ethyl-N-(4-methoxybenzyl)benzenesulphonamide
Vzorec: C₁₆H₁₉NO₃S
Storage Temperature: Ambient
MDL Number: MFCD09878415
CAS číslo: 1000339-36-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR