Tisk…

3-(4-Bromo-1H-pyrazol-1-yl)propan-1-ol

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Bromo-1-(3-hydroxypropyl)pyrazole

OR59395-1G OR59395-5G
APOSOR59395-1GEA 2440 CZK
APOSOR59395-1G APOSOR59395-5G
3-(4-Bromo-1H-pyrazol-1-yl)propan-1-ol
3-(4-Bromo-1H-pyrazol-1-yl)propan-1-ol
Vzorec: C₆H₉BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD06804869
CAS číslo: 925180-06-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR