Tisk…

2-Chloro-6-(1-pyrrolidinyl)pyrazine

Dodavatel: Apollo Scientific
Varování

OR59394-1G OR59394-5G
APOSOR59394-1GEA 3740 CZK
APOSOR59394-1G APOSOR59394-5G
2-Chloro-6-(1-pyrrolidinyl)pyrazine
2-Chloro-6-(1-pyrrolidinyl)pyrazine
Vzorec: C₈H₁₀ClN₃
MW: 183,64 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09864953
CAS číslo: 1000339-30-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR