Tisk…

2-Bromo-6-ethoxypyridine 95%

Dodavatel: Apollo Scientific
Varování

OR59382-1G OR59382-250MG
APOSOR59382-1GEA 1820 CZK
APOSOR59382-1G APOSOR59382-250MG
2-Bromo-6-ethoxypyridine 95%
2-Bromo-6-ethoxypyridine
Vzorec: C₇H₈BrNO
MW: 202.05
Storage Temperature: Ambient
MDL Number: MFCD00234958
CAS číslo: 4645-11-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR