2-Chloro-6-(ethylamino)pyrazine 98%
Dodavatel: Apollo Scientific
OR59339-1G
OR59339-5G
APOSOR59339-1GEA
5390
CZK
APOSOR59339-1G
APOSOR59339-5G
2-Chloro-6-(ethylamino)pyrazine 98%
2-Chloro-6-(ethylamino)pyrazine
Vzorec:
C₈H₁₂ClN₃ Bod tání: 94…96 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09878382 CAS číslo: 957065-84-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...