Tisk…

2-Morpholin-4-ylmethylphenylamine

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2-(Morpholin-4-ylmethyl)aniline

OR5922-1G OR5922-5G
APOSOR5922-1GEA 6290 CZK
APOSOR5922-1G APOSOR5922-5G
2-Morpholin-4-ylmethylphenylamine
2-Morpholin-4-ylmethylphenylamine
Vzorec: C₁₁H₁₆N₂O
MW: 192.26 g/mol
Bod tání: 69.5…71.5 °C
Storage Temperature: Ambient
MDL Number: MFCD04114505
CAS číslo: 95539-61-0
UN: 1759
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR