[3-(3-Chlorophenoxy)propyl]amine 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR5919-1G
OR5919-250MG
APOSOR5919-1GEA
8440
CZK
APOSOR5919-1G
APOSOR5919-250MG
[3-(3-Chlorophenoxy)propyl]amine 95%
[3-(3-Chlorophenoxy)propyl]amine
Vzorec:
C₉H₁₂ClNO MW: 185,66 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06800437 CAS číslo: 116753-50-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...