Tisk…

[2-(3-Chlorophenoxy)ethyl]amine 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2-(3-Chlorophenoxy)ethylamine

OR5917-1G OR5917-250MG
APOSOR5917-1GEA 5390 CZK
APOSOR5917-1G APOSOR5917-250MG
[2-(3-Chlorophenoxy)ethyl]amine 97%
[2-(3-Chlorophenoxy)ethyl]amine
Vzorec: C₈H₁₀ClNO
MW: 171,63 g/mol
Bod varu: 148…150 °C/16mmHg
Storage Temperature: Ambient
MDL Number: MFCD00052976
CAS číslo: 6488-00-2
UN: 1760
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR