4-(4,6-Dimethoxypyrimidin-2-yl)benzoic acid 95+%
Dodavatel: Apollo Scientific
OR5887-1G
OR5887-5G
OR5887-10G
APOSOR5887-1GEA
5960
CZK
APOSOR5887-1G
APOSOR5887-5G
APOSOR5887-10G
4-(4,6-Dimethoxypyrimidin-2-yl)benzoic acid 95+%
4-(4,6-Dimethoxypyrimidin-2-yl)benzoic acid
Vzorec:
C₁₃H₁₂N₂O₄ Bod tání: 284…286 °C Storage Temperature: Ambient |
MDL Number:
MFCD01568125 CAS číslo: 386715-40-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...