Tisk…

Indole-3-carboxylic acid 98%

Dodavatel: Apollo Scientific
Varování

OR5873-100G OR5873-25G OR5873-500G
APOSOR5873-100GEA 1080 CZK
APOSOR5873-100G APOSOR5873-25G APOSOR5873-500G
Indole-3-carboxylic acid 98%
Indole-3-carboxylic acid
Vzorec: C₉H₇NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD00005624
CAS číslo: 771-50-6
EINECS: 212-231-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR