(S)-(+)-Prolinol 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(S)-(+)-2-Hydroxymethylpyrrolidine, (2S)-pyrrolidin-2-ylmethanol, L(+)-Prolinol, S(+)-2-Pyrrolidinemethanol, L-Prolinol
OR5872-100G
OR5872-1G
OR5872-500G
APOSOR5872-100GEA
4070
CZK
APOSOR5872-100G
APOSOR5872-1G
APOSOR5872-500G
(S)-(+)-Prolinol 98%
(S)-(+)-Prolinol
Vzorec:
C₅H₁₁NO MW: 101,15 g/mol Bod varu: 74…76 °C/2mmHg Hustota: 1,025 g/cm³ (25 °C) Bod vzplanutí: 86 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005255 CAS číslo: 23356-96-9 EINECS: 245-605-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...