3-[4-(Methylsulphonyl)phenyl]-(2E)propenoic acid 95+%
Dodavatel: Apollo Scientific
|
Varování
|
OR5863-1G
OR5863-250MG
OR5863-5G
APOSOR5863-1GEA
3230
CZK
APOSOR5863-1G
APOSOR5863-250MG
APOSOR5863-5G
3-[4-(Methylsulphonyl)phenyl]-(2E)propenoic acid 95+%
3-[4-(Methylsulphonyl)phenyl]-(2E)propenoic acid
Vzorec:
C₁₀H₁₀O₄S MW: 226,25 g/mol Bod tání: 293…295 °C Storage Temperature: Ambient |
MDL Number:
MFCD00203126 CAS číslo: 5345-30-2 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...