Tisk…

(S)-Methyl-2-amino-4-methylpentanoate hydrochloride

Dodavatel: Apollo Scientific

Synonyms: H-Leu-OMe·HCl, H-Leu-OMe.HCl, L-Leucine methyl ester hydrochloride

OR5853-100G OR5853-25G
APOSOR5853-100GEA 787 CZK
APOSOR5853-100G APOSOR5853-25G
(S)-Methyl-2-amino-4-methylpentanoate hydrochloride
(S)-Methyl-2-amino-4-methylpentanoate hydrochloride
Vzorec: C₇H₁₅NO₂·HCl
MW: 181,7 g/mol
Bod tání: 145…155 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012494
CAS číslo: 7517-19-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR