5-Bromo-2-iodopyrimidine
Dodavatel: Apollo Scientific
|
Varování
|
OR5851-25G
OR5851-5G
APOSOR5851-25GEA
1370
CZK
APOSOR5851-25G
APOSOR5851-5G
5-Bromo-2-iodopyrimidine
5-Bromo-2-iodopyrimidine
Vzorec:
C₄H₂BrIN₂ MW: 284,88 g/mol Bod tání: 99…103 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01318111 CAS číslo: 183438-24-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...