Tisk…

tert-Butylchlorodiphenylsilane

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: tert-Butyldiphenylchlorosilane, tert-Butyl-diphenylchlorosilane

OR5837-100G OR5837-25G OR5837-500G
APOSOR5837-100GEA 1450 CZK
APOSOR5837-100G APOSOR5837-25G APOSOR5837-500G
tert-Butylchlorodiphenylsilane
tert-Butylchlorodiphenylsilane
Vzorec: C₁₆H₁₉ClSi
Bod varu: 90…92 °C/0,01mmHg
Hustota: 1,057 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000497
CAS číslo: 58479-61-1
EINECS: 261-282-0
UN: 2987
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR