Tisk…

3-Bromo-2-cyano-6-methylpyridine 95%

Dodavatel: Apollo Scientific

Synonyms: 3-Bromo-6-methylpicolinonitrile, 3-Bromo-6-methylpyridine-2-carbonitrile

OR5836-1G OR5836-5G
APOSOR5836-1GEA 5200 CZK
APOSOR5836-1G APOSOR5836-5G
3-Bromo-2-cyano-6-methylpyridine 95%
3-Bromo-2-cyano-6-methylpyridine
Vzorec: C₇H₅BrN₂
MW: 197,0300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08059560
CAS číslo: 717843-48-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR