3-Amino-5-nitrophenylboronic acid pinacol ester 97%
Dodavatel: Apollo Scientific
Synonyms:
3-Nitro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
OR5819-1G
OR5819-250MG
APOSOR5819-1GEA
19280
CZK
APOSOR5819-1G
APOSOR5819-250MG
3-Amino-5-nitrophenylboronic acid pinacol ester 97%
3-Amino-5-nitrophenylboronic acid pinacol ester
Vzorec:
C₁₂H₁₇BN₂O₄ MW: 264.09 Storage Temperature: Refrigerator |
MDL Number:
MFCD07363740 CAS číslo: 871329-51-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...