Tisk…

3-Chloro-4-(tert-butylcarbamoyl)phenylboronic acid 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-Chloro-4-(tert-butylcarbamoyl)benzeneboronic acid, 3-Chloro-4-(N-tert-butylcarbamoyl)phenylboronic acid

OR5818-1G OR5818-250MG OR5818-5G
APOSOR5818-1GEA 6450 CZK
APOSOR5818-1G APOSOR5818-250MG APOSOR5818-5G
3-Chloro-4-(tert-butylcarbamoyl)phenylboronic acid 95%
3-Chloro-4-(tert-butylcarbamoyl)phenylboronic acid
Vzorec: C₁₁H₁₅BClNO₃
MW: 255,51 g/mol
Bod tání: 130…134 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07363771
CAS číslo: 850589-46-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR