Tisk…

N-Benzyl-3-bromo-4-methylbenzenesulphonamide 98%

Dodavatel: Apollo Scientific

OR5810-1G OR5810-5G
APOSOR5810-1GEA 4860 CZK
APOSOR5810-1G APOSOR5810-5G
N-Benzyl-3-bromo-4-methylbenzenesulphonamide 98%
N-Benzyl-3-bromo-4-methylbenzenesulphonamide
Vzorec: C₁₄H₁₄BrNO₂S
Bod tání: 98…101 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07363832
CAS číslo: 850429-67-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR