Tisk…

N-Methyl-3-nitrobenzenesulphonamide 98%

Dodavatel: Apollo Scientific

OR5808-1G OR5808-5G
APOSOR5808-1GEA 2070 CZK
APOSOR5808-1G APOSOR5808-5G
N-Methyl-3-nitrobenzenesulphonamide 98%
N-Methyl-3-nitrobenzenesulphonamide
Vzorec: C₇H₈N₂O₄S
Bod tání: 110…114 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00474673
CAS číslo: 58955-78-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR