Tisk…

Tetrachloroterephthalic acid

Dodavatel: Apollo Scientific
Varování

OR5794-10G OR5794-1G OR5794-5G OR5794-25G
APOSOR5794-10GEA 11180 CZK
APOSOR5794-10G APOSOR5794-1G APOSOR5794-5G APOSOR5794-25G
Tetrachloroterephthalic acid
Tetrachloroterephthalic acid
Vzorec: C₈H₂Cl₄O₄
Storage Temperature: Ambient
CAS číslo: 2136-79-0

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR