Tisk…

2-Methoxy-4-methyl-6-(methylamino)-1,3,5-triazine

Dodavatel: Apollo Scientific

Synonyms: 4-Methoxy-N,6-dimethyl-1,3,5-triazin-2-amine

OR5792-100G OR5792-500G
APOSOR5792-100GEA 910 CZK
APOSOR5792-100G APOSOR5792-500G
2-Methoxy-4-methyl-6-(methylamino)-1,3,5-triazine
2-Methoxy-4-methyl-6-(methylamino)-1,3,5-triazine
Vzorec: C₆H₁₀N₄O
MW: 154.17 g/mol
Bod tání: 162…166 °C
Storage Temperature: Ambient
MDL Number: MFCD00585858
CAS číslo: 5248-39-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR