Tisk…

Tetrabromothiophene 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: Perbromothiophene

OR5778-100G OR5778-25G OR5778-500G
APOSOR5778-100GEA 3940 CZK
APOSOR5778-100G APOSOR5778-25G APOSOR5778-500G
Tetrabromothiophene 98%
Tetrabromothiophene
Vzorec: C₄Br₄S
Bod varu: 324…327 °C
Bod tání: 113…118 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005419
CAS číslo: 3958-03-0
EINECS: 223-554-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR