2-Acetamido-5-bromoisonicotinic acid 96%
Dodavatel: Apollo Scientific
|
Varování
|
OR5772-1G
OR5772-5G
APOSOR5772-1GEA
6220
CZK
APOSOR5772-1G
APOSOR5772-5G
2-Acetamido-5-bromoisonicotinic acid 96%
2-Acetamido-5-bromoisonicotinic acid
Vzorec:
C₈H₇BrN₂O₃ Bod tání: 258…262 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD07363802 CAS číslo: 871269-03-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...