2-Hydroxy-5-nitronicotinic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
5-Nitro-2-oxo-3-pyridinecarboxylic Acid
OR5764-1G
OR5764-25G
OR5764-5G
APOSOR5764-1GEA
622
CZK
APOSOR5764-1G
APOSOR5764-25G
APOSOR5764-5G
2-Hydroxy-5-nitronicotinic acid 98%
2-Hydroxy-5-nitronicotinic acid
Vzorec:
C₆H₄N₂O₅ MW: 184,11 g/mol Bod tání: 244…248 °C Storage Temperature: Ambient |
MDL Number:
MFCD05669267 CAS číslo: 6854-07-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...