4,4-Dimethylcyclohexa-1,5-dienylboronic acid monosodium salt 96%
Dodavatel: Apollo Scientific
Synonyms:
4,4-Dimethylcyclohexa-1,5-diene-1-boronic acid monosodium salt
OR5762-500MG
OR5762-250MG
APOSOR5762-500MGEA
0
CZK
APOSOR5762-500MG
APOSOR5762-250MG
4,4-Dimethylcyclohexa-1,5-dienylboronic acid monosodium salt 96%
4,4-Dimethylcyclohexa-1,5-dienylboronic acid monosodium salt
Vzorec:
C₈H₁₂BNaO₂ Bod tání: 300…302 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD07363759 CAS číslo: 871329-70-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...