2-Chloro-3-quinolinecarboxylic acid 95%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Chloroquinoline-3-carboxylic acid
OR5757-5G
OR5757-250MG
APOSOR5757-5GEA
5200
CZK
APOSOR5757-5G
APOSOR5757-250MG
2-Chloro-3-quinolinecarboxylic acid 95%
2-Chloro-3-quinolinecarboxylic acid
Vzorec:
C₁₀H₆ClNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD03030039 CAS číslo: 73776-25-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...