2-Iodopyridine 98%
Dodavatel: Apollo Scientific
|
Varování
|
OR5750-100G
OR5750-25G
APOSOR5750-100GEA
6450
CZK
APOSOR5750-100G
APOSOR5750-25G
2-Iodopyridine 98%
2-Iodopyridine
Vzorec:
C₅H₄IN MW: 205 g/mol Bod varu: 52 °C/0,9mmHg Hustota: 1,928 g/cm³ (25 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00464928 CAS číslo: 5029-67-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...