2-Amino-6-chloropyridine 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
6-Chloro-2-pyridylamine, 6-Chloropyridin-2-amine
OR5749-100G
OR5749-25G
APOSOR5749-100GEA
2570
CZK
APOSOR5749-100G
APOSOR5749-25G
2-Amino-6-chloropyridine 98%
2-Amino-6-chloropyridine
Vzorec:
C₅H₅ClN₂ MW: 128,56 g/mol Bod tání: 69…73 °C Storage Temperature: Ambient |
MDL Number:
MFCD00234068 CAS číslo: 45644-21-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...