3-(1,2,4-Triazol-3-yl)carbamoyl)phenylboronic acid 95%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(3-((1H-1,2,4-Triazol-3-yl)carbamoyl)phenyl)boronic acid
OR5719-1G
OR5719-250MG
APOSOR5719-1GEA
6950
CZK
APOSOR5719-1G
APOSOR5719-250MG
3-(1,2,4-Triazol-3-yl)carbamoyl)phenylboronic acid 95%
3-(1,2,4-Triazol-3-yl)carbamoyl)phenylboronic acid
Vzorec:
C₉H₉BN₄O₃ Bod tání: 230…236 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD07363798 CAS číslo: 871333-05-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...