Tisk…

3-((4,5-Dihydrothiazol-2-yl)carbamoyl)phenylboronic acid 97%

Dodavatel: Apollo Scientific

Synonyms: (3-((4,5-Dihydrothiazol-2-yl)carbamoyl)phenyl)boronic acid

OR5717-1G OR5717-5G
APOSOR5717-1GEA 5720 CZK
APOSOR5717-1G APOSOR5717-5G
3-((4,5-Dihydrothiazol-2-yl)carbamoyl)phenylboronic acid 97%
3-((4,5-Dihydrothiazol-2-yl)carbamoyl)phenylboronic acid
Vzorec: C₁₀H₁₁BN₂O₃S
MW: 250.0800 g/mol
Bod tání: 204…240 °C
Storage Temperature: Ambient
MDL Number: MFCD07363799
CAS číslo: 871333-06-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR